Anti-TSPAN17 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00026262-B01P
103342-248EA 798.46 USD
103342-248
Anti-TSPAN17 Mouse Polyclonal Antibody
Antibodies
The TSPAN17 Antibody from Novus Biologicals is a mouse polyclonal antibody to TSPAN17. This antibody reacts with human. The TSPAN17 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TSPAN17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR