Anti-TSPAN1 Mouse Monoclonal Antibody [clone: 5G11]

Supplier: Novus Biologicals
H00010103-M01
103338-146EA 399 USD
103338-146
Anti-TSPAN1 Mouse Monoclonal Antibody [clone: 5G11]
Antibodies
The TSPAN1 Antibody (5G11) from Novus Biologicals is a mouse monoclonal antibody to TSPAN1. This antibody reacts with human. The TSPAN1 Antibody (5G11) has been validated for the following applications: ELISA.

Type: Primary
Antigen: TSPAN1
Clonality: Monoclonal
Clone: 5G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG Mix Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR