Anti-TSLP Mouse Monoclonal Antibody (Biotin) [clone: 210H1.04]

Supplier: Novus Biologicals
DDX0804B
103319-042EA 734.58 USD
103319-042
Anti-TSLP Mouse Monoclonal Antibody (Biotin) [clone: 210H1.04]
Antibodies
The TSLP Antibody (210H1.04) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to TSLP. This antibody reacts with human. The TSLP Antibody (210H1.04) [Biotin] has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TSLP
Clonality: Monoclonal
Clone: 210H1.04
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR