Anti-TSLP Mouse Monoclonal Antibody [clone: 209A2.03]

Supplier: Novus Biologicals
DDX0801P-100
103319-040EA 578.89 USD
103319-040
Anti-TSLP Mouse Monoclonal Antibody [clone: 209A2.03]
Antibodies
The TSLP Antibody (209A2.03) from Novus Biologicals is a mouse monoclonal antibody to TSLP. This antibody reacts with human. The TSLP Antibody (209A2.03) has been validated for the following applications: ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: TSLP
Clonality: Monoclonal
Clone: 209A2.03
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR