Anti-TSHZ2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-88303
103271-612EA 734.58 USD
103271-612
Anti-TSHZ2 Rabbit Polyclonal Antibody
Antibodies
The TSHZ2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TSHZ2. This antibody reacts with human. The TSHZ2 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TSHZ2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR