Anti-TSC22/TSC22D1 Mouse Monoclonal Antibody [clone: 1G7]

Supplier: Novus Biologicals
H00008848-M01
103335-762EA 721.54 USD
103335-762
Anti-TSC22/TSC22D1 Mouse Monoclonal Antibody [clone: 1G7]
Antibodies
The TSC22 / TSC22D1 Antibody (1G7) from Novus Biologicals is a mouse monoclonal antibody to TSC22 / TSC22D1. This antibody reacts with human. The TSC22 / TSC22D1 Antibody (1G7) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: TSC22/TSC22D1
Clonality: Monoclonal
Clone: 1G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR