Anti-TSC22/TSC22D1 Mouse Polyclonal Antibody

H00008848-B01P
103335-758EA 583.08 USD
103335-758
Anti-TSC22/TSC22D1 Mouse Polyclonal Antibody
The TSC22 / TSC22D1 Antibody from Novus Biologicals is a mouse polyclonal antibody to TSC22 / TSC22D1. This antibody reacts with human. The TSC22 / TSC22D1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: TSC22/TSC22D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR