Anti-TSC22/TSC22D1 Rabbit Polyclonal Antibody

H00008848-D01P
103335-760EA 627.69 USD
103335-760
Anti-TSC22/TSC22D1 Rabbit Polyclonal Antibody
The TSC22 / TSC22D1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TSC22 / TSC22D1. This antibody reacts with human. The TSC22 / TSC22D1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TSC22/TSC22D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR