Anti-TSC1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
24920002-0.1MG
103316-714EA 349 USD
103316-714
Anti-TSC1 Rabbit Polyclonal Antibody
Antibodies
The TSC1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TSC1. This antibody reacts with human. The TSC1 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TSC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR