Anti-TRXR2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-92499
103284-446EA 734.58 USD
103284-446
Anti-TRXR2 Rabbit Polyclonal Antibody
Antibodies
The Thioredoxin-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Thioredoxin-2. This antibody reacts with human. The Thioredoxin-2 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TRXR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR