Anti-TRXR1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00007296-D01P
103333-550EA 429 USD
103333-550
Anti-TRXR1 Rabbit Polyclonal Antibody
Antibodies
The Thioredoxin Reductase 1 / TRXR1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Thioredoxin Reductase 1 / TRXR1. This antibody reacts with human. The Thioredoxin Reductase 1 / TRXR1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: TRXR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR