Anti-TRPC6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20737
102712-344EA 635.51 USD
102712-344
Anti-TRPC6 Rabbit Polyclonal Antibody
Antibodies
The TRPC6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRPC6. This antibody reacts with human. The TRPC6 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: TRPC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR