Anti-TRPC5 Mouse Monoclonal Antibody [clone: 1C8]

Supplier: Novus Biologicals
H00007224-M10
103333-396EA 798.46 USD
103333-396
Anti-TRPC5 Mouse Monoclonal Antibody [clone: 1C8]
Antibodies
The TRPC5 Antibody (1C8) from Novus Biologicals is a mouse monoclonal antibody to TRPC5. This antibody reacts with human. The TRPC5 Antibody (1C8) has been validated for the following applications: Western Blot, ELISA.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: TRPC5
Clonality: Monoclonal
Clone: 1C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR