Anti-TRPC5 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00007224-B01P
103333-394EA 798.46 USD
103333-394
Anti-TRPC5 Mouse Polyclonal Antibody
Antibodies
The TRPC5 Antibody from Novus Biologicals is a mouse polyclonal antibody to TRPC5. This antibody reacts with human. The TRPC5 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TRPC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR