Anti-TRPC3 Rabbit Polyclonal Antibody

NBP1-86266
103272-690EA 607.69 USD
103272-690
Anti-TRPC3 Rabbit Polyclonal Antibody
The TRPC3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRPC3. This antibody reacts with human. The TRPC3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TRPC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR