Anti-TROY/TNFRSF19 Mouse Monoclonal Antibody (DyLight 350) [clone: 933214]

Supplier: Novus Biologicals
MAB1548UV
103293-764EA 614.15 USD
103293-764
Anti-TROY/TNFRSF19 Mouse Monoclonal Antibody (DyLight 350) [clone: 933214]
Antibodies
The TROY / TNFRSF19 Antibody (933214) [DyLight 350] from Novus Biologicals is a mouse monoclonal antibody to TROY / TNFRSF19. This antibody reacts with human. The TROY / TNFRSF19 Antibody (933214) [DyLight 350] has been validated for the following applications: Flow Cytometry.

Type: Primary
Antigen: TROY/TNFRSF19
Clonality: Monoclonal
Clone: 933214
Conjugation: DyLight 350
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR