Anti-TROVE2 Mouse Monoclonal Antibody [clone: 2E10.]

Supplier: Novus Biologicals
H00006738-M04
103310-784EA 798.46 USD
103310-784
Anti-TROVE2 Mouse Monoclonal Antibody [clone: 2E10.]
Antibodies
The TROVE2 Antibody (2E10.) from Novus Biologicals is a mouse monoclonal antibody to TROVE2. This antibody reacts with human. The TROVE2 Antibody (2E10.) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TROVE2
Clonality: Monoclonal
Clone: 2E10.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR