Anti-TRMU Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-87923
103274-408EA 734.58 USD
103274-408
Anti-TRMU Rabbit Polyclonal Antibody
Antibodies
The TRMU Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRMU. This antibody reacts with human. The TRMU Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TRMU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR