Anti-TRMT61B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-83640
103267-930EA 734.58 USD
103267-930
Anti-TRMT61B Rabbit Polyclonal Antibody
Antibodies
The TRMT61B Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRMT61B. This antibody reacts with human. The TRMT61B Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TRMT61B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR