Anti-TRMT61A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20728
102712-326EA 635.51 USD
102712-326
Anti-TRMT61A Rabbit Polyclonal Antibody
Antibodies
The TRMT61A Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRMT61A. This antibody reacts with human. The TRMT61A Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TRMT61A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR