Anti-TrkB Mouse Polyclonal Antibody

H00004915-B01P
103331-952EA 553.85 USD
103331-952
Anti-TrkB Mouse Polyclonal Antibody
The TrkB Antibody from Novus Biologicals is a mouse polyclonal antibody to TrkB. This antibody reacts with human. The TrkB Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TrkB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR