Anti-TrkB Mouse Monoclonal Antibody [clone: 3D12]

H00004915-M02
103331-958EA 553.85 USD
103331-958
Anti-TrkB Mouse Monoclonal Antibody [clone: 3D12]
The TrkB Antibody (3D12) from Novus Biologicals is a mouse monoclonal antibody to TrkB. This antibody reacts with human. The TrkB Antibody (3D12) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TrkB
Clonality: Monoclonal
Clone: 3D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR