Anti-TRIP6 Mouse Monoclonal Antibody [clone: 4B7]

Supplier: Novus Biologicals
H00007205-M04
103333-390EA 798.46 USD
103333-390
Anti-TRIP6 Mouse Monoclonal Antibody [clone: 4B7]
Antibodies
The TRIP6 Antibody (4B7) from Novus Biologicals is a mouse monoclonal antibody to TRIP6. This antibody reacts with human. The TRIP6 Antibody (4B7) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: TRIP6
Clonality: Monoclonal
Clone: 4B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR