Anti-TRIML1 Mouse Monoclonal Antibody [clone: 4F8]

Supplier: Novus Biologicals
H00339976-M01
103348-270EA 798.46 USD
103348-270
Anti-TRIML1 Mouse Monoclonal Antibody [clone: 4F8]
Antibodies
The TRIML1 Antibody (4F8) from Novus Biologicals is a mouse monoclonal antibody to TRIML1. This antibody reacts with human. The TRIML1 Antibody (4F8) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TRIML1
Clonality: Monoclonal
Clone: 4F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR