Anti-TRIM9 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00114088-D01P
103346-134EA 844.62 USD
103346-134
Anti-TRIM9 Rabbit Polyclonal Antibody
Antibodies
The TRIM9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRIM9. This antibody reacts with human, mouse. The TRIM9 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TRIM9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR