Anti-TRIM65 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00201292-B01P
103347-716EA 798.46 USD
103347-716
Anti-TRIM65 Mouse Polyclonal Antibody
Antibodies
The TRIM65 Antibody from Novus Biologicals is a mouse polyclonal antibody to TRIM65. This antibody reacts with human. The TRIM65 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TRIM65
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR