Anti-TRIM61 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-13485
103281-096EA 734.58 USD
103281-096
Anti-TRIM61 Rabbit Polyclonal Antibody
Antibodies
The TRIM61 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRIM61. This antibody reacts with human. The TRIM61 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TRIM61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR