Anti-TRIM47 Mouse Monoclonal Antibody [clone: 3C8]

Supplier: Novus Biologicals
H00091107-M02
103345-772EA 798.46 USD
103345-772
Anti-TRIM47 Mouse Monoclonal Antibody [clone: 3C8]
Antibodies
The TRIM47 Antibody (3C8) from Novus Biologicals is a mouse monoclonal antibody to TRIM47. This antibody reacts with human. The TRIM47 Antibody (3C8) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: TRIM47
Clonality: Monoclonal
Clone: 3C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR