Anti-TRIM42 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20718
102712-306EA 635.51 USD
102712-306
Anti-TRIM42 Rabbit Polyclonal Antibody
Antibodies
The TRIM42 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRIM42. This antibody reacts with human. The TRIM42 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TRIM42
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR