Anti-TRIM40 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00135644-B01P
103346-892EA 798.46 USD
103346-892
Anti-TRIM40 Mouse Polyclonal Antibody
Antibodies
The TRIM40 Antibody from Novus Biologicals is a mouse polyclonal antibody to TRIM40. This antibody reacts with human. The TRIM40 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: TRIM40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR