Anti-TRIM37 Mouse Monoclonal Antibody [clone: 2D11]

Supplier: Novus Biologicals
H00004591-M01
103331-236EA 798.46 USD
103331-236
Anti-TRIM37 Mouse Monoclonal Antibody [clone: 2D11]
Antibodies
The TRIM37 Antibody (2D11) from Novus Biologicals is a mouse monoclonal antibody to TRIM37. This antibody reacts with human. The TRIM37 Antibody (2D11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TRIM37
Clonality: Monoclonal
Clone: 2D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR