Anti-TRIM35 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00023087-D01P
103340-980EA 429 USD
103340-980
Anti-TRIM35 Rabbit Polyclonal Antibody
Antibodies
The TRIM35 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRIM35. This antibody reacts with human. The TRIM35 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: TRIM35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR