Anti-TRIM32 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00022954-B01P
103340-866EA 798.46 USD
103340-866
Anti-TRIM32 Mouse Polyclonal Antibody
Antibodies
The TRIM32 Antibody from Novus Biologicals is a mouse polyclonal antibody to TRIM32. This antibody reacts with human. The TRIM32 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TRIM32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR