Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRIM31 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00011074-B01P
103340-088
Anti-TRIM31 Mouse Polyclonal Antibody
The TRIM31 Antibody from Novus Biologicals is a mouse polyclonal antibody to TRIM31. This antibody reacts with human. The TRIM31 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TRIM31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty