Anti-TRIM25 Mouse Monoclonal Antibody [clone: 5C3]

Supplier: Novus Biologicals
H00007706-M03
103334-292EA 798.46 USD
103334-292
Anti-TRIM25 Mouse Monoclonal Antibody [clone: 5C3]
Antibodies
The TRIM25 Antibody (5C3) from Novus Biologicals is a mouse monoclonal antibody to TRIM25. This antibody reacts with human. The TRIM25 Antibody (5C3) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TRIM25
Clonality: Monoclonal
Clone: 5C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR