Anti-TRIM25 Mouse Monoclonal Antibody [clone: 2B12]

Supplier: Novus Biologicals
H00007706-M01
103334-288EA 399 USD
103334-288
Anti-TRIM25 Mouse Monoclonal Antibody [clone: 2B12]
Antibodies
The TRIM25 Antibody (2B12) from Novus Biologicals is a mouse monoclonal antibody to TRIM25. This antibody reacts with human. The TRIM25 Antibody (2B12) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TRIM25
Clonality: Monoclonal
Clone: 2B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR