Anti-TRIM25 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00007706-B01P
103334-284EA 399 USD
103334-284
Anti-TRIM25 Mouse Polyclonal Antibody
Antibodies
The TRIM25 Antibody from Novus Biologicals is a mouse polyclonal antibody to TRIM25. This antibody reacts with human. The TRIM25 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: TRIM25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR