Anti-TRIM23 Mouse Monoclonal Antibody [clone: 2G4]

Supplier: Novus Biologicals
H00000373-M12
103322-054EA 798.46 USD
103322-054
Anti-TRIM23 Mouse Monoclonal Antibody [clone: 2G4]
Antibodies
The TRIM23 Antibody (2G4) from Novus Biologicals is a mouse monoclonal antibody to TRIM23. This antibody reacts with human. The TRIM23 Antibody (2G4) has been validated for the following applications: ELISA.

Type: Primary
Antigen: TRIM23
Clonality: Monoclonal
Clone: 2G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR