Anti-TRIM21/SSA1 Mouse Monoclonal Antibody [clone: 2C9]

Supplier: Novus Biologicals
H00006737-M01
103310-776EA 798.46 USD
103310-776
Anti-TRIM21/SSA1 Mouse Monoclonal Antibody [clone: 2C9]
Antibodies
The TRIM21 / SSA1 Antibody (2C9) from Novus Biologicals is a mouse monoclonal antibody to TRIM21 / SSA1. This antibody reacts with human. The TRIM21 / SSA1 Antibody (2C9) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TRIM21/SSA1
Clonality: Monoclonal
Clone: 2C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR