Anti-TRIM17 Mouse Monoclonal Antibody [clone: 2D3]

Supplier: Novus Biologicals
H00051127-M03
103343-702EA 399 USD
103343-702
Anti-TRIM17 Mouse Monoclonal Antibody [clone: 2D3]
Antibodies
The TRIM17 Antibody (2D3) from Novus Biologicals is a mouse monoclonal antibody to TRIM17. This antibody reacts with human. The TRIM17 Antibody (2D3) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: TRIM17
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR