Anti-TRIM16 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-84921
103263-446EA 734.58 USD
103263-446
Anti-TRIM16 Rabbit Polyclonal Antibody
Antibodies
The TRIM16 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRIM16. This antibody reacts with human. The TRIM16 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TRIM16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR