Anti-TRIM15 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00089870-B01P
103345-644EA 798.46 USD
103345-644
Anti-TRIM15 Mouse Polyclonal Antibody
Antibodies
The TRIM15 Antibody from Novus Biologicals is a mouse polyclonal antibody to TRIM15. This antibody reacts with human. The TRIM15 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TRIM15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR