Anti-TRIB1 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-27557
103409-288EA 564.74 USD
103409-288
Anti-TRIB1 Goat Polyclonal Antibody
Antibodies
The TRIB1 Antibody from Novus Biologicals is a goat polyclonal antibody to TRIB1. This antibody reacts with mouse. The TRIB1 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.

Type: Primary
Antigen: TRIB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR