Anti-TRF-2 Mouse Monoclonal Antibody (DyLight 755) [clone: 4A794.15]

Supplier: Novus Biologicals
NB100-56506IR
103359-360EA 688 USD
103359-360
Anti-TRF-2 Mouse Monoclonal Antibody (DyLight 755) [clone: 4A794.15]
Antibodies
The TRF-2 Antibody (4A794.15) [DyLight 755] from Novus Biologicals is a mouse monoclonal antibody to TRF-2. This antibody reacts with human, mouse, rat. The TRF-2 Antibody (4A794.15) [DyLight 755] has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: TRF-2
Clonality: Monoclonal
Clone: 4A794.15
Conjugation: DyLight 755
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR