Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRF-2 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 4A794.15]

Supplier: Novus Biologicals
NB100-56506H
103359-358
Anti-TRF-2 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 4A794.15]
The TRF-2 Antibody (4A794.15) [HRP] from Novus Biologicals is a mouse monoclonal antibody to TRF-2. This antibody reacts with human, mouse, rat. The TRF-2 Antibody (4A794.15) [HRP] has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: TRF-2
Clonality: Monoclonal
Clone: 4A794.15
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty