Anti-SUMO2 Mouse Monoclonal Antibody [clone: 2H8]

H00006613-M06
103310-412EA 583.08 USD
103310-412
Anti-SUMO2 Mouse Monoclonal Antibody [clone: 2H8]
The SUMO2 Antibody (2H8) from Novus Biologicals is a mouse monoclonal antibody to SUMO2. This antibody reacts with human. The SUMO2 Antibody (2H8) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SUMO2
Clonality: Monoclonal
Clone: 2H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR