Anti-SUMO2 Rabbit Polyclonal Antibody

H00006613-D01P
103310-408EA 627.69 USD
103310-408
Anti-SUMO2 Rabbit Polyclonal Antibody
The SUMO2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SUMO2. This antibody reacts with human. The SUMO2 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SUMO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR