Anti-SUMO1 Mouse Monoclonal Antibody [clone: 3D7]

Supplier: Novus Biologicals
H00007341-M03
103333-632EA 798.46 USD
103333-632
Anti-SUMO1 Mouse Monoclonal Antibody [clone: 3D7]
Antibodies
The SUMO1 Antibody (3D7) from Novus Biologicals is a mouse monoclonal antibody to SUMO1. This antibody reacts with human. The SUMO1 Antibody (3D7) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SUMO1
Clonality: Monoclonal
Clone: 3D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR