Anti-SUGT1 Mouse Polyclonal Antibody

H00010910-A03
103339-778EA 436.92 USD
103339-778
Anti-SUGT1 Mouse Polyclonal Antibody
The SUGT1 Antibody from Novus Biologicals is a mouse polyclonal antibody to SUGT1. This antibody reacts with human. The SUGT1 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SUGT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR