Anti-SUB1 Mouse Monoclonal Antibody [clone: 3A1]

Supplier: Novus Biologicals
H00010923-M11
103339-818EA 798.46 USD
103339-818
Anti-SUB1 Mouse Monoclonal Antibody [clone: 3A1]
Antibodies
The SUB1 Antibody (3A1) from Novus Biologicals is a mouse monoclonal antibody to SUB1. This antibody reacts with human. The SUB1 Antibody (3A1) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SUB1
Clonality: Monoclonal
Clone: 3A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR